Learn More
Nefazodone hydrochloride, MedChemExpress
Product Code. 16478698
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16478698
Brand: Medchem Express HYB1396
MedChemExpress Nefazodone hydrochloride (BMY-13754) is a potent and selective 5HT2A (Ki=5.8 nM) antagonist with moderate inhibition of 5-HT and noradrenaline uptake (IC50 of 290 and 300 nM, respectively). Nefazodone hydrochloride is a phenylpiperazine antidepressant with less alpha-adrenergic blocking activity.
Nefazodone hydrochloride (BMY-13754) is a potent and selective 5HT2A (Ki=5.8 nM) antagonist with moderate inhibition of 5-HT and noradrenaline uptake (IC50 of 290 and 300 nM, respectively). Nefazodone hydrochloride is a phenylpiperazine antidepressant with less alpha-adrenergic blocking activity.
Specifications
Chemical Name or Material Nefazodone hydrochloride
CAS 82752-99-6
Color White
Purity Grade Notes Research
Molecular Formula C25H33Cl2N5O2
Quantity 10 mg
Synonym BMY-13754 MJ-13754-1
Solubility Information DMSO : 50 mg/mL (98.72 mM; Need ultrasonic) ∣H2O : 2 mg/mL (3.95 mM; Need ultrasonic)
SMILES O=C1N(CCCN2CCN(C3=CC=CC(Cl)=C3)CC2)N=C(CC)N1CCOC4=CC=CC=C4.[H]Cl
Molecular Weight (g/mol) 506.47
Formula Weight 506.47
Percent Purity 99.02%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.