Learn More
NEDD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13993-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody works well in WB, IHC, IP and IF application.
Neural precursor cell expressed developmentally down-regulated protein 1 (NEDD1) is required for mitosis progression. NEDD1 promotes the nucleation of microtubules from the spindle.Specifications
| NEDD1 | |
| Polyclonal | |
| Unconjugated | |
| NEDD1 | |
| GCP WD, NEDD 1, NEDD1, Protein NEDD1, TUBGCP7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 121441, 17997, 299730 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P33215, Q8NHV4 | |
| NEDD1 | |
| NEDD1 Fusion Protein Ag5077 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |