Description
NDUFV1 is the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.The NDUFV1 gene encodes the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.
Specifications
Specifications
| Antigen | NDUFV1 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | NDUFV1 |
| Gene Accession No. | P49821, Q91YT0 |
| Gene Alias | CI 51kD, Complex I 51kD, NDUFV1, UQOR1 |
| Gene Symbols | Ndufv1 |
| Show More |