missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDUFS2 Monoclonal Antibody (OTI2G2)
GREENER_CHOICE

Product Code. 15916134
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15916134 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15916134 Supplier Invitrogen™ Supplier No. MA526534

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NDUFS2 Monoclonal Antibody for IHC (P), Western Blot

The protein encoded by this gene is a core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase. Mammalian mitochondrial complex I is composed of at least 43 different subunits, 7 of which are encoded by the mitochondrial genome, and the rest are the products of nuclear genes. The iron-sulfur protein fraction of complex I is made up of 7 subunits, including this gene product. Complex I catalyzes the NADH oxidation with concomitant ubiquinone reduction and proton ejection out of the mitochondria. Mutations in this gene are associated with mitochondrial complex I deficiency. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFS2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2G2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 1% BSA and 0.02% sodium azide; pH 7.3
Gene Ndufs2
Gene Accession No. O75306
Gene Alias AL033311; CI-49; CI-49kD; comp; complex 1, mitochondrial respiratory chain, 49-KD subunit; complex I 49kDa subunit; complex I-49kD; EMBL:AAH82067.1}; NADH dehydrogenase (ubiquinone) Fe-S protein 2; NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial; NADH:ubiquinone oxidoreductase core subunit S2; NADH-coenzyme Q reductase; NADH-ubiquinone oxidoreductase 49 kDa subunit; NADH-ubiquinone oxidoreductase NDUFS2 subunit; Ndufs2; ndufs2 {ECO:0000312; unnamed protein product
Gene Symbols Ndufs2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 205-463 of NDUFS2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4720
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.