Learn More
NDUFS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16810164 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16810164 20 μL 20µL
16800164 150 μL 150µL
2 options
Product Code. 16810164
Brand: Proteintech 124441AP20UL
Rabbit Polyclonal Antibody
The protein encoded by this gene belongs to the complex I 75 kDa subunit family. Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. This protein is the largest subunit of complex I and it is a component of the iron-sulfur fragment of the enzyme. It may form part of the active site crevice where NADH is oxidized. Mutations in this gene are associated with complex I deficiency. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
Antigen NDUFS1
Applications Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry
Classification Polyclonal
Concentration 0.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene NDUFS1
Gene Accession No. P28331, Q66HF1, Q91VD9
Gene Alias CI 75Kd, Complex I 75kD, NDUFS1, PRO1304
Gene Symbols NDUFS1
Host Species Rabbit
Immunogen NDUFS1 Fusion Protein Ag3135
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227197, 301458, 4719
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG