Learn More
Abnova™ NDUFB8 (Human) Recombinant Protein
Product Code. 16128231
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16128231
Brand: Abnova™ H00004714P01.10ug
Human NDUFB8 full-length ORF ( AAH00466, 1 a.a. - 186 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MAVARAGVLGVQWLQRASRNVMPLGARTASHMTKDMFPGPYPRTPEERAAAAKKYNMRVEDYEPYPDDGMGYGDYPKLPDRSQHERDPWYSWDQPGLRLNWGEPMHWHLDMYNRNRVDTSPTPVSWHVMCMQLFGFLAFMIFMCWVGDVYPVYQPVGPKQYPYNNLYLERGGDPSKEPERVVHYEI
Specifications
Accession Number AAH00466
Gene ID (Entrez) 4714
Name NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 8, 19kDa
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias ASHI, CI-ASHI
Gene Symbol NDUFB8
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer