missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFB7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14912-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a subunit of the multisubunit NADH:ubiquinone oxidoreductase. Mammalian complex I is composed of 45 different subunits. It is located at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.Specifications
NDUFB7 | |
Polyclonal | |
Unconjugated | |
NDUFB7 | |
B18, Cell adhesion protein SQM1, CI B18, Complex I B18, NDUFB7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361385, 4713, 66916 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P17568, Q9CR61 | |
Ndufb7 | |
NDUFB7 Fusion Protein Ag6709 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |