missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NDUFAB1 (Human) Recombinant Protein
Human NDUFAB1 full-length ORF ( AAH58920, 1 a.a. - 156 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004706-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MASRVLSAYVSRLPAAFAPLPRVRMLAVARPLSTALCSAGTQTRLGTLQPALVLAQVPGRVTQLCRQYSDMPPLTLEGIQDRVLYVLKLYDKIDPEKLSVNSHFMKDLGLDSLDQVEIIMAMEDEFGFEIPYIDAEKLMCPQEIVDYIADKKDVYE
Specifications
AAH58920 | |
NADH dehydrogenase (ubiquinone) 1, alpha/beta subcomplex, 1, 8kDa | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NDUFAB1 | |
GST |
4706 | |
Wheat germ expression system | |
25 μg | |
ACP, FASN2A, MGC65095, SDAP | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |