missing translation for 'onlineSavingsMsg'
Learn More

NDUFA5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16856324 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16856324 20 μL 20µL
16846324 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16856324

Brand: Proteintech 166401AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.

Specifications

Antigen NDUFA5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NDUFA5
Gene Accession No. Q16718, Q63362, Q9CPP6
Gene Alias B13, CI 13KD B, Complex I 13kD B, Complex I subunit B13, NDUFA5, NUFM, UQOR13
Gene Symbols NDUFA5
Host Species Rabbit
Immunogen NDUFA5 Fusion Protein Ag9995
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25488, 4698, 68202
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.