Description
NDUFA4L2 is a protein coding gene.
Specifications
Specifications
| Antigen | NDUFA4L2 |
| Applications | Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | NDUFA4L2 |
| Gene Accession No. | Q9NRX3 |
| Gene Alias | NDUFA4L2, NUOMS |
| Gene Symbols | Ndufa4l2 |
| Show More |