missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ NDUFA12 Recombinant Protein

Brand:  Abnova™ H00055967-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16188546

  • 6245.00 NOK / 25µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH05936
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
41.58
8
Glutathione Sepharose 4 Fast Flow
25 ug
MELVQVLKRGLQQITGHGGLRGYLRVFFRTNDAKVGTLVGEDKYGNKYYEDNKQFFGRHRWVVYTTEMNGKNTFWDVDGSMVPPEWHRWLHSMTDDPPTTKPLAARKFIWTNHKFNVTGTPEQYVPYSTTRKKIQEWIPPSTPYK
B17.2/DAP13
NDUFA12
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
55967
NDUFA12 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NDUFA12
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers