missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NBR1 Mouse anti-Human, Mouse, Rat, Clone: 1F2B2, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | NBR1 |
|---|---|
| Clone | 1F2B2 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17689726
|
Proteintech
68062-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17679726
|
Proteintech
68062-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NBR1 Monoclonal antibody apecifically detetects NBR1 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot.Specifications
| NBR1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| BC009808 | |
| NBR1 | |
| IgG1 | |
| Protein G purification |
| 1F2B2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| 4077 | |
| NBR1 fusion protein Ag8652 | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title