Learn More
Nav1.7 inhibitor, MedChemExpress
Product Code. 16496376
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16496376
Brand: Medchem Express HY13985
MedChemExpress Nav1.7 inhibitor (compound II), a sulfonamide, is a potent Nav1.7 inhibitor. Nav1.7 inhibitor has the potential for a wide range of disorders, particularly pain.
Nav1.7 inhibitor (compound II), a sulfonamide, is a potent Nav1.7 inhibitor. Nav1.7 inhibitor has the potential for a wide range of disorders, particularly pain.
Specifications
Chemical Name or Material Nav1.7 inhibitor
CAS 1355631-24-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H11Cl3FNO4S
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (234.37 mM)
SMILES O=C(NS(=O)(C)=O)C1=CC(Cl)=C(COC2=CC=C(Cl)C(Cl)=C2)C=C1F
Molecular Weight (g/mol) 426.67
Formula Weight 426.67
Percent Purity 98.81%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.