missing translation for 'onlineSavingsMsg'
Learn More

Nav1.7 inhibitor, MedChemExpress

Product Code. 16485948
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485948

Brand: Medchem Express HY13985

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Nav1.7 inhibitor (compound II), a sulfonamide, is a potent Nav1.7 inhibitor. Nav1.7 inhibitor has the potential for a wide range of disorders, particularly pain.

Nav1.7 inhibitor (compound II), a sulfonamide, is a potent Nav1.7 inhibitor. Nav1.7 inhibitor has the potential for a wide range of disorders, particularly pain.

Specifications

Chemical Name or Material Nav1.7 inhibitor
CAS 1355631-24-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H11Cl3FNO4S
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (234.37 mM)
SMILES O=C(NS(=O)(C)=O)C1=CC(Cl)=C(COC2=CC=C(Cl)C(Cl)=C2)C=C1F
Molecular Weight (g/mol) 426.67
Formula Weight 426.67
Percent Purity 98.81%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.