missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nav1.5 Antibody (4G8:1G7), Novus Biologicals™
Mouse Monoclonal Antibody
1975.00 NOK
Specifications
| Antigen | Nav1.5 |
|---|---|
| Clone | 4G8:1G7 |
| Concentration | 1 mg/ml |
| Dilution | Western Blot 2 μg/mL, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000, CyTOF-ready |
| Applications | Western Blot |
Description
Nav1.5 Monoclonal specifically detects Nav1.5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Radioimmunoassay, CyTOF-ready.Specifications
| Nav1.5 | |
| 1 mg/ml | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| cardiac sodium channel alpha subunit, cardiac tetrodotoxin-insensitive voltage-dependent sodium channel alpha subunit, CDCD2VF1, HB1sodium channel, voltage-gated, type V, alpha (long QT syndrome 3), HB2, HBBD, HH1CMD1E, ICCD, IVF, LQT3SSS1CMPD2, Nav1.5, PFHB1, Sodium channel protein cardiac muscle subunit alpha, sodium channel protein type 5 subunit alpha, sodium channel protein type V alpha subunit, Sodium channel protein type V subunit alpha, sodium channel, voltage-gated, type V, alpha subunit, Voltage-gated sodium channel subunit alpha Nav1.5 | |
| SCN5A | |
| IgG1 | |
| Affinity Purified | |
| Nav1.5 (4G8:1G7). |
| 4G8:1G7 | |
| Western Blot 2 μg/mL, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| 6331 | |
| Retinoblastoma tissue membrane fraction | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title