Learn More
Deutsch Neumann™ Bulbs for Smaller Pipettes
Product Code. 10591832
Change view
Click to view available options
Volume (Metric):
2 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10591832 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10591832 Supplier Deutsch Neumann™ Supplier No. 4600004
  • ONLINE EXCLUSIVE PRICE
    7.33 NOK
    6.06 NOK / Pack of 1
    Save 1.27 NOK
    17% Off
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
Excellent for use with Pasteur pipettes. Deutsch Neumann™ Bulbs for Smaller Pipettes are manufactured from natural rubber and silicon.
  • Red color
  • Provides excellent suction
Specifications
Material Natural Rubber
Type Bulb
Volume (Metric) 2 mL
For Use With (Equipment) Smaller Pipettes
Quantity 1 Each