Learn More
Deutsch Neumann™ The Flip™ Safety Pipette Fillers
Product Code. 10436511
Change view
Click to view available options
Color:
Green
Red
Unit Size:
Each
Product Code. Color unitSize
10436511 Green Each
10473194 Red Each
2 options
Product Code. 10436511
Brand: Deutsch Neumann™ 4710021
Only null left
Add to basket
Add to basket
Reduces the risks of pipetting when handling toxic, corrosive, bacteria contaminated or bad smelling liquids. Deutsch Neumann™ The Flip™ Safety Pipette Fillers are manufactured from natural rubber.
  • Only two operating points
  • Evacuate via the new automatic valve
  • Available in a range of colors
  • One-handed design is the simplest safety pipette filler available
  • Ability to clean inside of bulb by removing patented valve and rinsing out
Specifications
Color Green
Material Natural Rubber
Type 'Flip' Safety Pipette Filler
Quantity 1 Each