Learn More
BRAND™ Natural Rubber Pipet Filler
Product Code. 10221601 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
10 mL
100 mL
Model:
Flip
Standard
Universal
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Model Unit Size
10221601 10 mL Standard Each
10363341 100 mL Universal Each
10110671 100 mL Flip Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10221601 Supplier BRAND™ Supplier No. 25300
  • ONLINE EXCLUSIVE PRICE
    207.00 NOK
    130.85 NOK / Pack of 1
    Save 76.15 NOK
    36% Off
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
Bulk Quote Request
Manufactured from 100% natural rubber. BRAND™ Natural Rubber Pipet Filler eliminates mouth pipetting and handling of hazardous material. Available in universal, standard, or flip model, with 2 or 3-valve options.
  • Manufactured from 100% natural rubber.
  • Eliminates mouth pipetting and handling of hazardous material.
  • Available in universal, standard, or flip model, with 2 or 3-valve options
  • 2-valve flip model enables one-handed operation.
TRUSTED_SUSTAINABILITY
Specifications
Description Valve A: Release air, Valve S: Filling, Valve E: Delivery
Material Natural Rubber
Model Standard
Volume (Metric) 10 mL
For Use With (Application) Filling pipets up to 10mL
Includes 3 valves
Quantity 1 Each