Learn More
Ansell Edmont™ Profil™ Plus 87-850 Series Yellow Natural Rubber Latex Immersion Gloves
Shop All Ansell Products
Click to view available options
Unit Size:
12 pairs
Size:
8
9
Description
- Specially treated to reduce the risk of allergy
- Excellent resistance to mechanical hazards and enhanced chemical resistance
- 100% cotton flocklining - soft, comfortable and absorbent
- Ideal for janitorial use and handling in wet conditions
Specifications
Specifications
| Back Material | Rubber Latex |
| Cuff Material | Natural Rubber Latex |
| Coating Material | Natural Rubber Latex |
| Latex Free | No |
| Product Type | Immersion Gloves |
| Style | 87-850 |
| Thickness (Metric) | 0.55 mm |
| Glove Cuff Color | Yellow |
| Glove Cuff Style | Pinked |
| Liner Material | Cotton Flocking |
| Show More |