Learn More
Deutsch Neumann™ Natural Rubber Double Bellows
Product Code. 10507951
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10507951 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10507951 Supplier Deutsch Neumann™ Supplier No. 4100003
  • ONLINE EXCLUSIVE PRICE
    542.00 NOK
    415.48 NOK / Pack of 1
    Save 126.52 NOK
    23% Off
View all promotions
Estimated Shipment: 23-11-2026
Use this natural rubber bellows for a variety of applications. Deutsch Neumann™ Natural Rubber Double Bellows feature inlet and outlet valves and a short piece of 3mm bore tubing connected to the outlet.
  • Double spray bellows  with net
  • Manufactured from black natural rubber
Specifications
Color Black
Material Natural Rubber
Type Double Blower