Learn More
Deutsch Neumann™ Rubber Balloon with Hose Connection
Product Code. 10418301
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
10418301 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10418301 Supplier Deutsch Neumann™ Supplier No. 4500002
Add to basket
Add to basket
Provides for control of air/gas entry or tread out of gas in instruments. Deutsch Neumann™ Rubber Balloon with Hose Connection is manufactured from latex natural rubber.
  • For rotary adapters
  • Suitable for mixture or collection and short-term storage of gases
Specifications
Material Natural Rubber
Type Balloon
For Use With (Equipment) For rotary evaporators