Learn More
Masterflex™ Natural Polypropylene Pipe Nipple Threaded Fittings
Product Code. 15139222
Change view
Click to view available options
Port Size:
1-1/2 x 11/32 x 21/32in.
1-29/32 x 13/16 x 1-5/16in.
1-7/8 x 19/32 x 1-1/32in.
Unit Size:
Pack of 10
Pack of 5
Product Code. Port Size unitSize
15139222 1-29/32 x 13/16 x 1-5/16in. Pack of 5
13566720 1-1/2 x 11/32 x 21/32in. Pack of 10
15257847 1-7/8 x 19/32 x 1-1/32in. Pack of 10
3 options
Product Code. 15139222
Brand: Masterflex™ 4061094
Only null left
Specifications
Material Polypropylene
Min. Operating Temperature (Metric) -17°C
Packaging Quantity 5 Pack
Port Size 1-29/32 x 13/16 x 1-5/16in.
Thread Size 3/4 in. MNPT
Max. Operating Temperature (Metric) 87°C
Product Type Pipe Nipple Threaded Fitting