Learn More
Kautex™ LDPE Wide Mouth Bottles Series 303
Product Code. 11962659
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
200 mL
250 mL
300 mL
500 mL
750 mL
1000 mL
1500 mL
2000 mL
Neck ID (Metric):
23.8 mm
30.8 mm
39.2 mm
50 mm
Unit Size:
Each
Pack of 10
Pack of 180
Pack of 20
Pack of 4
Pack of 5
Pack of 50
Pack of 90
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Neck ID (Metric) unitSize
11962659 2000 mL 50 mm Each
11324455 50 mL 23.8 mm Pack of 50
11899684 100 mL 23.8 mm Pack of 20
10607303 100 mL 23.8 mm Each
12684940 200 mL 30.8 mm Pack of 10
15241609 250 mL 30.8 mm Pack of 10
11910517 300 mL 30.8 mm Pack of 10
17878107 300 mL 30.8 mm Pack of 180
11344455 500 mL 39.2 mm Pack of 10
15251609 750 mL 39.2 mm Pack of 5
17848097 750 mL 39.2 mm Pack of 90
11361984 1000 mL 50 mm Pack of 4
11371984 1500 mL 50 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 11962659 Supplier Kautex™ Supplier No. 2000770537
Add to basket
Edge
Add to basket
Designed with a wide opening for easy filling and cleaning. Kautex™ Wide Mouth Bottles are ideal for liquids, pastes and granules.
  • Manufactured from low density polyethylene (LDPE) for flexibility and toughness
  • Natural translucent color, round bottles
  • Easy to fill and clean
  • Ideal for dispensing, sending samples, adhesives, transportation and storage 
  • Includes leakproof screw closure
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 2000 mL
For Use With (Application) Dispensing, Sending Samples, Mailing, Transport, Storage
Form Round
Includes Closure
Material LDPE
Neck ID (Metric) 50 mm
Replacement Closure Screw Closure LDPE
Quantity 1 Ea.
Graduated No
Height (Metric) 247 mm
Product Type Wide Mouth Bottle
Closure Type Screw Cap