Learn More
Ark Plas Products™ Premium Grade Natural Kynar Threaded Elbow Barbed Fitting
An excellent option for connecting tubing in the laboratory, with a pressure rating of up to 125 P.S.I. without clamps.
Brand:  Ark Plas Products™ MEX21-DF0
Additional Details : Weight : 0.04680kg
Product Code. 11841160
  • 1055.00 NOK / Pack of 10
Estimated Shipment: 30-08-2024
to see stock.
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Description
Description
  • Manufactured to the most exacting standards
  • No parting line on the barb
  • Molded from virgin, lot-traceable materials only
  • Made with critical use applications in mind
Specifications
Specifications
Elbow Barbed Threaded Connector
10 Pack
1/4-18 NPT Male Thread
Kynar
1/4 in. Barb
Natural