Learn More
Ark Plas Products™ Premium Grade Natural Kynar Threaded Elbow Barbed Fitting
An excellent option for connecting tubing in the laboratory, with a pressure rating of up to 125 P.S.I. without clamps.
Brand: Ark Plas Products™ MEX21-DF0
Additional Details : Weight : 0.04680kg
Description
- Manufactured to the most exacting standards
- No parting line on the barb
- Molded from virgin, lot-traceable materials only
- Made with critical use applications in mind
Specifications
Elbow Barbed Threaded Connector | |
10 Pack | |
1/4-18 NPT Male Thread |
Kynar | |
1/4 in. Barb | |
Natural |