Learn More
Reitenspiess™ Test Tube Brush with Wool Tip
Product Code. 10097900
Change view
Click to view available options
Length (Metric):
220 mm
270 mm
280 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10097900 270 mm Each
10692931 280 mm Each
10031074 220 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10097900 Supplier Reitenspiess™ Supplier No. 50300334
  • ONLINE EXCLUSIVE PRICE
    45.06 NOK
    25.82 NOK / Pack of 1
    Save 19.24 NOK
    42% Off
Estimated Shipment: 26-11-2026
Clean your test tubes using the Reitenspiess™ Test Tube Brush with Wool Tip. Use a test tube brush to clean your test tubes and other laboratory ware.
  • Natural bristles
  • Wool Tip
  • Sterile
Specifications
Product Type Test Tube Brush
Sterility Sterile
For Use With (Application) For cleaning, disinfection, sterilization.
Length (Metric) 270 mm
Quantity 1 Ea.