Learn More
Reitenspiess™ Test Tube Brush with Wool Tip
Product Code. 10031074
Change view
Click to view available options
Length (Metric):
220 mm
270 mm
280 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10031074 220 mm Each
10692931 280 mm Each
10097900 270 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10031074 Supplier Reitenspiess™ Supplier No. 12220301
Outlet Corner
Use promo code 28080
View all promotions
In Stock
Add to basket
Edge
Add to basket
Clean your test tubes using the Reitenspiess™ Test Tube Brush with Wool Tip. Use a test tube brush to clean your test tubes and other laboratory ware.
  • Natural bristles
  • Wool Tip
  • Sterile
Specifications
Length (Metric) 220 mm
Length (Metric) Bristle 80 mm
Length (Metric) Brush 105 mm
Quantity 1 Ea.