Learn More
Scharlab NATRIUMTETRABORAT-10-HYDR.PS 1KG
Brand:  Scharlab SO07051000
NATRIUMTETRABORAT-10-HYDR.PS 1KG
Product Code. 11338574
  • ONLINE EXCLUSIVE PRICE
    1013.25 NOK
    754.47 NOK / 1kg
    Save 258.78 NOK
    25% Off
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • NATRIUMTETRABORAT-10-HYDR.PS 1KG
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : disodium tetraboratedecahydrate