Learn More
Scharlab NATRIUMTETRABORAT-10-HYDR.PA 1KG
Brand:  Scharlab SO07071000
NATRIUMTETRABORAT-10-HYDR.PA 1KG
Product Code. 11358574
  • 1496.25 NOK / 1kg
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • di-Sodium tetraborate decahydrate, reagent grade, ACS, ISO
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .