Learn More
SCS Bulk Chemicals NATRIUMDEOXYKOLAT 1KG SPEC PLASTICPACK,MAX3% WATER
Brand: SCS Bulk Chemicals 218590000
NATRIUMDEOXYKOLAT 1KG SPEC PLASTICPACK,MAX3% WATER
This item is not returnable.
View return policy
Description
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Deoxycholic acid, sodium salt
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
Hazard Statement
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.