Learn More
Scharlab NATRIUMAZID PA 100GSegregation: Away from heavy metals and their salt
Brand: Scharlab SO00910100
NATRIUMAZID PA 100GSegregation: Away from heavy metals and their salt
Safety and Handling
- Sodium azide, extra pure, Reag. Ph Eur
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H300-Fatal if swallowed.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P273-Avoid release to the environment.
- P264-Wash thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to .