Learn More
Thermo Scientific™ National Unassembled Standard Opening Vial, Cap and Septum Convenience Kits
Description
Keep all items handy and dust-free with Thermo Scientific™ National Unassembled Vial, Cap and Septum Convenience Kits.
- Two-compartment, see-through benchtop storage tray
- Caps with septa are not attached to vials (packaged in plastic bags)
Includes:
100 vials and 100 caps with pre-assembled septa
Specifications
Specifications
| Closure Color | Black |
| Color (Seal) | Red/White/Red |
| Description | Vials: Clear Glass; Closure: Black; Septa: Red PTFE/White Silicone/Red PTFE |
| Material (Liner) | PTFE/Silicone |
| Color | Clear |
| Material | Glass |
| Septum | PTFE/Silicone/PTFE |
| Marking Spot | No |