Learn More
Thermo Scientific™ National Unassembled Standard Opening Vial, Cap and Septum Convenience Kits
Product Code. 12460108
Change view
Click to view available options
Marking Spot:
No
Yes
Septum:
PTFE/Silicone
PTFE/Silicone/PTFE
Color:
Clear
Color (Seal):
Red/White
Red/White/Red
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Marking Spot Septum Color Color (Seal) unitSize
12460108 No PTFE/Silicone/PTFE Clear Red/White/Red Pack of 100
12450128 Yes PTFE/Silicone Clear Red/White Pack of 100
12400168 No PTFE/Silicone Clear Red/White Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12460108 Supplier Thermo Scientific™ Supplier No. C401316
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Keep all items handy and dust-free with Thermo Scientific™ National Unassembled Vial, Cap and Septum Convenience Kits.
Keep all items handy and dust-free with Thermo Scientific™ National Unassembled Vial, Cap and Septum Convenience Kits.
  • Two-compartment, see-through benchtop storage tray
  • Caps with septa are not attached to vials (packaged in plastic bags)
Includes: 
100 vials and 100 caps with pre-assembled septa
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Black
Color (Seal) Red/White/Red
Description Vials: Clear Glass; Closure: Black; Septa: Red PTFE/White Silicone/Red PTFE
Material (Liner) PTFE/Silicone
Color Clear
Material Glass
Septum PTFE/Silicone/PTFE
Marking Spot No