Learn More
Thermo Scientific™ National Target™ Unassembled Vial, Cap and Septum Convenience Kits
Description
Keep vials, caps and septa within easy reach and dust free with Thermo Scientific™ National Target Unassembled Convenience Kits.
- Two-compartment benchtop storage tray
- I-D™ vials feature write-on patch with graduations
- All items remain handy and dust-free
- Caps with septa are not attached to vials (packed in separate polybag)
Includes:
100 vials and 100 caps with pre-assembled septa
Specifications
Specifications
| Closure Color | Light Blue |
| Color (Seal) | Red/White/Red |
| Description | Amber Glass; Light Blue Cap, Red PTFE/White Silicone/Red PTFE Septum; 2mL |
| Color | Amber |
| Material | Glass |
| Pre-slit | No |
| Volume (Metric) | 2 mL |
| Septum | PTFE/Silicone/PTFE |
| Marking Spot | No |