Learn More
Thermo Scientific™ National Target™ Snap-It™ Kits
Description
Thermo Scientific™ National Target Snap-It Convenience Kits provide vials and Snap-it caps with prefitted septa in storage trays.
- 2mL,12 × 32mm vials
- Choice of vial and preassembled seals with septa
- Packaged in two compartment benchtop storage trays
- Include matched quantities of vials and Snap-It caps with prefitted septa
- All items remain dust-free
Specifications
Specifications
| Closure Color | Clear |
| Description | Clear Glass ID; Septum: PTFE/Silicone, Pre-Slit |
| Color | Clear |
| Material | PTFE, Silicone |
| Pre-slit | Yes |
| Volume (Metric) | 2 mL |
| Septum | PTFE/Silicone, Pre-slit |
| Closure Material | PTFE, Silicone |
| Marking Spot | Yes |