Learn More
Thermo Scientific™ National Target™ Snap Cap Wide-Opening Vials
Click to view available options
Type:
Micro-V™ Amber Glass Snap-It Microsampling Vial
Micro-V™ Clear Glass Snap-It Microsampling Vial
Unit Size:
Pack of 100
Description
Thermo Scientific™ National Target Snap Cap Wide-Opening Vials are easier to fill and provide a larger target area for the autosampler needle.
- 2mL, 12 x 32mm Crimp/Snap Ring Vials
- Fit rotating or robotic arm autosamplers
- I-D™ vials feature write-on patch with graduations at 0.5, 1.0 and 1.5mL
- Target Snap-It™ vials can be used with snap caps or aluminum crimp seal closures
- Fit most autosamplers that accommodate the 12 × 32mm vial format
- Superior quality 33 expansion borosilicate clear glass (Type 1, Class A) or 51A amber glass
- Uniformly flat bottom for security with inserts
Specifications
Specifications
| Description | Micro-V Clear Glass Snap-It Microsampling Vial |
| Volume (Metric) | 15 μL |
| Volume (Metric) Residual | 15 μL |
| Type | Micro-V™ Clear Glass Snap-It Microsampling Vial |