Learn More
Thermo Scientific™ National Target LoVial™ Crimp-Top Vials—Wide-Opening 2mL, 12 x 32mm
Product Code. 12463718
Click to view available options
Volume (Metric):
2 mL
2.5 mL
Volume (Metric) Working:
1.5 mL
1.8 mL
2.3 mL
Unit Size:
Pack of 100
Pack of 1000
Type:
Kimshield Deactivated Amber Glass I-D Vial
Kimshield Deactivated Clear Glass I-D Vial
Product Code. 12463718
Brand: Thermo Scientific™ C40111240
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ National Target LoVial Crimp-Top Vials are designed with a wide-opening for easier filling.
Thermo Scientific™ National Target LoVial Crimp-Top Vials are designed with a wide-opening for easier filling.
  • Fit most autosamplers that accommodate the 12 × 32mm vial format
  • Borosilicate clear glass Type 1, Class A or 51A amber glass
  • Uniformly flat bottom for security with inserts
  • I-D™ vials feature write-on patch with graduations
  • Target Colorband™ crimp top vials feature color-coded graduations and patch
  • Vials with an 11mm crimp finish require aluminum seal closures
  • Crimp/snap vials can be used with either 11mm aluminum seals or Snap-It™ Snap Caps
TRUSTED_SUSTAINABILITY
Specifications
Description 2.5mL Clear Glass Crimp-Top Vial—40mm Vial Height
Volume (Metric) 2.5 mL
Volume (Metric) Working 2.3 mL