Learn More
Thermo Scientific™ National Target™ Assembled Vial, Cap and Septum Kits
Product Code. 12463628
Click to view available options
Marking Spot:
No
Yes
Closure Color:
Black
Light Blue
Septum:
PTFE/Silicone, Pre-slit
Red PTFE/White Silicone
Color:
Amber
Clear
Unit Size:
Pack of 100
Product Code. 12463628
Brand: Thermo Scientific™ C401017BLK
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
  • Assembled kits include 100 vials with pre-attached caps and septa
  • Packaged in convenient vial trays with clear covers or in polybags
  • Vial trays are stackable for easy storage
  • Vial trays hold vials upright for easy filling and maintaining sample order
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Black
Description Clear Glass; Black cap; PTFE/Silicone septum
Material (Liner) PTFE/Silicone
Color Clear
Material Glass
Septum Red PTFE/White Silicone
Marking Spot No