Learn More
Thermo Scientific™ National Target™ Assembled Vial, Cap and Septum Kits
Description
Thermo Scientific™ National Target Assembled Vial Kits include vials with pre-attached caps and septa, all packaged in convenient trays or polybags.
- Assembled kits include 100 vials with pre-attached caps and septa
- Packaged in convenient vial trays with clear covers or in polybags
- Vial trays are stackable for easy storage
- Vial trays hold vials upright for easy filling and maintaining sample order
Specifications
Specifications
| Closure Color | Light Blue |
| Color (Seal) | Red/White |
| Description | Amber Glass I-D; Light Blue Cap; Red PTFE/White Silicone septum |
| Material (Liner) | PTFE/Silicone |
| Color | Amber |
| Material | Glass |
| Septum | Red PTFE/White Silicone |
| Marking Spot | Yes |