Learn More
Thermo Scientific™ National Silanized Sample Vials
Product Code. 12488686
Change view
Click to view available options
Volume (Metric):
8 mL
12 mL
40 mL
Unit Size:
Pack of 100
Pack of 200
Product Code. Volume (Metric) unitSize
12488686 40 mL Pack of 100
12468686 12 mL Pack of 200
12478686 8 mL Pack of 200
3 options
Product Code. 12488686
Brand: Thermo Scientific™ B7999S6
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ National Silanized Sample Vials are constructed of superior quality borosilicate clear or amber glass.
Thermo Scientific™ National Silanized Sample Vials are constructed of superior quality borosilicate clear or amber glass.
TRUSTED_SUSTAINABILITY
Specifications
Description Clear; 40mL
Diameter (Metric) Outer 28 mm
Material Glass
Volume (Metric) 40 mL
Closure Size 24-400
Height (Metric) 95 mm