Learn More
Thermo Scientific™ National Microvolume Inserts for 8 x 40mm Shell Vials
Product Code. 12420208
Click to view available options
Volume (Metric):
275 μL
300 μL
Style:
Precision Point Tip
Pulled Point Tip
Unit Size:
Pack of 100
Product Code. 12420208
Brand: Thermo Scientific™ C4015536
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ National Microvolume Inserts are available in pulled point or precision point design to suit various application needs.
Thermo Scientific™ National Microvolume Inserts are available in pulled point or precision point design to suit various application needs.
  • Polyspring™ inserts feature a polyethylene spring to center the bottom of the insert and to provide a cushion against needle contact
  • Precision point inserts permit maximum sample extraction
  • Use metal spring with inserts that lack the Polyspring
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 275 μL
Style Pulled Point Tip
Product Line National
For Use With (Equipment) 8 x 40 mm Shell Vials