Learn More
Thermo Scientific™ Manual Crimpers and Decrimpers for Cleanrooms
Product Code. 15220379
Change view
Click to view available options
Size:
11 mm
13 mm
20 mm
Type:
Manual Crimper
Manual Decrimper
Unit Size:
Each
Product Code. Size Type unitSize
15220379 20 mm Manual Decrimper Each
15200379 11 mm Manual Crimper Each
15277298 13 mm Manual Crimper Each
15267298 11 mm Manual Decrimper Each
15210379 13 mm Manual Decrimper Each
15217308 20 mm - Each
6 options
Product Code. 15220379
Brand: Thermo Scientific™ C4020102SS
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use stainless-steel Thermo Scientific™ National Manual Crimpers and Decrimpers.
Crimpers provide a reproducible, secure vial closure; decrimpers allow easy removal of aluminum seals without breakage.
  • Stainless steel construction
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
No. per Pack 1 Ea.
Type Manual Decrimper
For Use With (Equipment) Aluminum Seal
Size 20 mm
For Use With (Application) Industrial Chromatography
Product Line National