Learn More
Thermo Scientific™ Manual Crimpers and Decrimpers for Cleanrooms
Product Code. 15217308
Change view
Click to view available options
Size:
11 mm
13 mm
20 mm
Unit Size:
Each
Type:
Manual Crimper
Manual Decrimper
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize Type
15217308 20 mm Each -
15200379 11 mm Each Manual Crimper
15277298 13 mm Each Manual Crimper
15267298 11 mm Each Manual Decrimper
15210379 13 mm Each Manual Decrimper
15220379 20 mm Each Manual Decrimper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15217308 Supplier Thermo Scientific™ Supplier No. C4020100SS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use stainless-steel Thermo Scientific™ National Manual Crimpers and Decrimpers.
Crimpers provide a reproducible, secure vial closure; decrimpers allow easy removal of aluminum seals without breakage.
  • Stainless steel construction
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Description Manual Crimper
No. per Pack 1 Ea.
For Use With (Equipment) Chromatography, Serum Vial
Size 20 mm
For Use With (Application) Industrial Chromatography
Product Line National