Learn More
Thermo Scientific™ National Economy Vials
Product Code. 11702197 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Amber
Clear
Product Type:
Clear
Crimp Top Vial
Screw Thread Vial
Snap cap
Volume (Metric) Working:
2 mL
Unit Size:
Pack of 1000
Product Code. Color Product Type Volume (Metric) Working unitSize
11702197 Amber Screw Thread Vial 2 mL Pack of 1000
11573830 Clear Crimp Top Vial 2 mL Pack of 1000
11583830 Amber Crimp Top Vial 2 mL Pack of 1000
11593830 Clear Clear 2 mL Pack of 1000
13287489 Amber Snap cap 2 mL Pack of 1000
10003873 Clear Screw Thread Vial 2 mL Pack of 1000
11563860 Clear Screw Thread Vial 2 mL Pack of 1000
11573860 Amber Screw Thread Vial 2 mL Pack of 1000
8 options
Product Code. 11702197
Brand: Thermo Scientific™ 60180715
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ National Economy Vials are offered as clear or amber, in a range of styles to meet specific application needs.
Thermo Scientific™ National Economy Vials are offered as clear or amber, in a range of styles to meet specific application needs.
TRUSTED_SUSTAINABILITY
Specifications
Description 60180-715
Color Amber
Volume (Metric) 2 mL
Volume (Metric) Working 2 mL
For Use With (Equipment) Thermo Scientific, Agilent, PE, Shimadzu, Varian
Product Type Screw Thread Vial