Learn More
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits
Product Code. 11911613
Click to view available options
Color:
Amber
Clear
Closure Type:
Phenolic
Polypropylene
Polypropylene, Flanged
Septum:
PTFE
PTFE/Silicone
PTFE/Silicone, Pre-slit
PTFE/Silicone/PTFE
Packaging Type:
Polybag
Storage Tray
Unit Size:
Pack of 100
Product Code. 11911613
Brand: Thermo Scientific™ C4013015A
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
  • Packaged in convenient vial trays with clear covers or in economical polybags
  • Vial trays are stackable for easy storage
  • All caps are manufactured from virgin polypropylene unless noted
Includes: 
100 vials with pre-attached caps and septa
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Black
Color (Seal) Red/White
Description Clear Glass; PP Black Cap; Red PTFE/White Silicone septum; in polybag
Color Clear
Material Glass, PTFE, Silicone
Volume (Metric) 2 mL
Septum PTFE/Silicone
Packaging Type Polybag
Closure Material Polypropylene
Closure Type Polypropylene
Closure Size 8-425