Learn More
Thermo Scientific™ National Assembled Screw Caps with Inserted Septa for 4mL Vials
Product Code. 12420178
Click to view available options
Color:
Black
Red
Material:
PTFE, Silicone, Polypropylene
Septum:
Red PTFE/White Silicone
White PTFE/Natural Silicone
Unit Size:
Pack of 100
Product Code. 12420178
Brand: Thermo Scientific™ C40151050A
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ National Assembled Screw Caps with Inserted Septa for 4mL Vials are designed to be used with any 13mm septa.
Thermo Scientific™ National Assembled Screw Caps with Inserted Septa for 4mL Vials are designed to be used with any 13mm septa.
TRUSTED_SUSTAINABILITY
Specifications
Description PP; Black; White PTFE/Natural Silicone
Color Black
Material PTFE, Silicone, Polypropylene
Septum White PTFE/Natural Silicone
Product Line National
For Use With (Equipment) 4 mL Vials