Learn More
Thermo Scientific™ 20mm Crimp Seals with Septa for Headspace Vials
Click to view available options
Septum:
Aluminum Foil/White High Temp. Silicone
Clear PTFE/gray butyl
SepCap Integral molded Polyethylene
Unit Size:
Pack of 100
Pack of 250
Type:
Crimp Cap
Pressure Release
Description
Minimize the risk of contamination with quick, convenient Thermo Scientific™ National 20mm Crimp Seals with Septa.
20mm snap cap with prefitted septa for headspace vials
- Seals with pre-inserted septa are convenient and minimize contamination due to handling
- Use magnetic seals with CTC/Leap™ Technologies, Gerstel™ and other magnetic transport autosamplers
- PurePak™ PTFE/Silicone seals are packed in glass jars to maintain purity up to the point of use
- Molded polyethylene SepCap™ is suitable for short-term storage or where repeated access to the vial is required
Ordering Information:
Septa and snap cap also sold separately.
Note:
SepCap is for short-term storage. Do not use SepCap at elevated temperatures.
Specifications
Specifications
| Description | SepCap Integral molded Polyethylene septa |
| Material | Polyethylene, Molded Polyethylene |
| Septum | SepCap Integral molded Polyethylene |
| Product Line | National |
| Closure Size | 20 mm |
| For Use With (Equipment) | Headspace Vials |