missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nateglinide

Product Code. p-7214432 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
16584364 1 g 1g
16594364 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 16584364

Brand: Thermo Scientific Chemicals 462980010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105816-04-4
Molecular Formula C19H27NO3
Molecular Weight (g/mol) 317.43
InChI Key CZCVBLROPSHDRZ-YESZJQIVSA-N
IUPAC Name (2R)-3-phenyl-2-{[(1S,3S)-3-(propan-2-yl)cyclohexyl]formamido}propanoic acid
SMILES CC(C)[C@H]1CCC[C@@H](C1)C(=O)N[C@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 129.0°C to 130.0°C
Quantity 1 g
Formula Weight 317.42
Chemical Name or Material Nateglinide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.