Learn More
Naspm trihydrochloride, MedChemExpress
Description
Naspm trihydrochloride (1-Naphthylacetyl spermine trihydrochloride), a synthetic analogue of Joro spider toxin, is a calcium permeable AMPA (CP-AMPA) receptors antagonist.
Specifications
Specifications
| Chemical Name or Material | Naspm trihydrochloride |
| Physical Form | Solid |
| CAS | 1049731-36-3 |
| Color | Pale Yellow |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H37Cl3N4O |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Synonym | 1-Naphthylacetyl spermine trihydrochloride |
| Solubility Information | H2O : 50 mg/mL (104.19 mM; Need ultrasonic) ∣DMSO : 6.4 mg/mL (13.34 mM; Need ultrasonic) |
| Show More |