missing translation for 'onlineSavingsMsg'
Learn More

Naspm trihydrochloride, MedChemExpress

Product Code. 16407947
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16407947 5 mg 5mg
16461437 50 mg 50mg
16415228 10 mg 10mg
16405906 100 mg 100mg
4 options
This item is not returnable. View return policy

Product Code. 16407947

Brand: Medchem Express HY12506A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Naspm trihydrochloride (1-Naphthylacetyl spermine trihydrochloride), a synthetic analogue of Joro spider toxin, is a calcium permeable AMPA (CP-AMPA) receptors antagonist.

Naspm trihydrochloride (1-Naphthylacetyl spermine trihydrochloride), a synthetic analogue of Joro spider toxin, is a calcium permeable AMPA (CP-AMPA) receptors antagonist.

Specifications

Chemical Name or Material Naspm trihydrochloride
Physical Form Solid
CAS 1049731-36-3
Color Pale Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H37Cl3N4O
For Use With (Application) Neuroscience-Neuromodulation
Synonym 1-Naphthylacetyl spermine trihydrochloride
Solubility Information H2O : 50 mg/mL (104.19 mM; Need ultrasonic) ∣DMSO : 6.4 mg/mL (13.34 mM; Need ultrasonic)
SMILES [H]Cl.[H]Cl.[H]Cl.O=C(NCCCNCCCCNCCCN)CC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 479.91
Formula Weight 479.91
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.