Learn More
Naspm trihydrochloride, MedChemExpress
Product Code. 16405906
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16405906
Brand: Medchem Express HY12506A
MedChemExpress Naspm trihydrochloride (1-Naphthylacetyl spermine trihydrochloride), a synthetic analogue of Joro spider toxin, is a calcium permeable AMPA (CP-AMPA) receptors antagonist.
Naspm trihydrochloride (1-Naphthylacetyl spermine trihydrochloride), a synthetic analogue of Joro spider toxin, is a calcium permeable AMPA (CP-AMPA) receptors antagonist.
Specifications
Chemical Name or Material Naspm trihydrochloride
Physical Form Solid
CAS 1049731-36-3
Color Pale Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C22H37Cl3N4O
For Use With (Application) Neuroscience-Neuromodulation
Synonym 1-Naphthylacetyl spermine trihydrochloride
Solubility Information H2O : 50 mg/mL (104.19 mM; Need ultrasonic) ∣DMSO : 6.4 mg/mL (13.34 mM; Need ultrasonic)
SMILES [H]Cl.[H]Cl.[H]Cl.O=C(NCCCNCCCCNCCCN)CC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 479.91
Formula Weight 479.91
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.